2,2,3,3,3-pentafluoro-1-(4-isopropoxy-2-methylphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

303829-1G 303829-500MG 303829-5G
77468-590EA 415.09 USD
77468-590 77481-274 77491-656
2,2,3,3,3-pentafluoro-1-(4-isopropoxy-2-methylphenyl)propan-1-one ≥95%
2,2,3,3,3-pentafluoro-1-(4-isopropoxy-2-methylphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₃H₁₃F₅O₂
MW: 296.2331 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892143
CAS Number: 2969328-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR