4-(4-Chlorophenoxy)-2-methylaniline ≥95%

Supplier: AOBChem USA

Warning

303801-1G 303801-500MG 303801-5G
77468-582EA 191.58 USD
77468-582 77481-266 77491-648
4-(4-Chlorophenoxy)-2-methylaniline ≥95%
4-(4-Chlorophenoxy)-2-methylaniline

Precaución: For research use only

Formula: C₁₃H₁₂ClNO
Storage Temperature: Ambient
MDL Number: MFCD08687440
CAS Number: 946743-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR