2-Chloro-2,2-difluoro-1-(2-methoxy-4-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

303785-1G 303785-500MG 303785-5G
77468-578EA 415.09 USD
77468-578 77481-262 77491-644
2-Chloro-2,2-difluoro-1-(2-methoxy-4-methylphenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(2-methoxy-4-methylphenyl)ethanone

Precaución: For research use only

Formula: C₁₀H₉ClF₂O₂
MW: 234.6270 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892118
CAS Number: 2969459-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR