Isopropyl(3-methyl-5-((3-methylbenzyl)oxy)phenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

303782-1G 303782-500MG 303782-5G
77468-574EA 415.09 USD
77468-574 77481-258 77491-640
Isopropyl(3-methyl-5-((3-methylbenzyl)oxy)phenyl)sulfane ≥95%
Isopropyl(3-methyl-5-((3-methylbenzyl)oxy)phenyl)sulfane

Precaución: For research use only

Formula: C₁₈H₂₂OS
Storage Temperature: Ambient
MDL Number: MFCD34892115
CAS Number: 2969459-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR