2-(3-Methyl-5-((3-methylbenzyl)oxy)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

303780-1G 303780-500MG 303780-5G
77468-570EA 415.09 USD
77468-570 77481-254 77491-636
2-(3-Methyl-5-((3-methylbenzyl)oxy)phenyl)propan-2-ol ≥95%
2-(3-Methyl-5-((3-methylbenzyl)oxy)phenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₈H₂₂O₂
MW: 270.3660 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892113
CAS Number: 2969475-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR