2-Ethyl-1-(3-methyl-5-((3-methylbenzyl)oxy)phenyl)butan-1-ol ≥95%

Supplier: AOBChem USA

Warning

303778-1G 303778-500MG 303778-5G
77468-566EA 415.09 USD
77468-566 77481-250 77491-632
2-Ethyl-1-(3-methyl-5-((3-methylbenzyl)oxy)phenyl)butan-1-ol ≥95%
2-Ethyl-1-(3-methyl-5-((3-methylbenzyl)oxy)phenyl)butan-1-ol

Precaución: For research use only

Formula: C₂₁H₂₈O₂
Storage Temperature: Ambient
MDL Number: MFCD34892111
CAS Number: 2969459-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR