1-Iodo-3-methyl-5-((3-methylbenzyl)oxy)benzene ≥95%

Supplier: AOBChem USA

Warning

303775-1G 303775-500MG 303775-5G
77468-562EA 415.09 USD
77468-562 77481-246 77491-628
1-Iodo-3-methyl-5-((3-methylbenzyl)oxy)benzene ≥95%
1-Iodo-3-methyl-5-((3-methylbenzyl)oxy)benzene

Precaución: For research use only

Formula: C₁₅H₁₅IO
MW: 338.1834 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892108
CAS Number: 2969459-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR