Triethyl(3-methyl-5-((3-methylbenzyl)oxy)phenyl)silane ≥95%

Supplier: AOBChem USA

Warning

303772-1G 303772-500MG 303772-5G
77468-558EA 415.09 USD
77468-558 77481-242 77491-624
Triethyl(3-methyl-5-((3-methylbenzyl)oxy)phenyl)silane ≥95%
Triethyl(3-methyl-5-((3-methylbenzyl)oxy)phenyl)silane

Precaución: For research use only

Formula: C₂₁H₃₀OSi
Storage Temperature: Ambient
MDL Number: MFCD34892105
CAS Number: 2969205-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR