2-Chloro-2,2-difluoro-1-(3-methyl-5-((3-methylbenzyl)oxy)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

303769-1G 303769-500MG 303769-5G
77468-554EA 415.09 USD
77468-554 77481-238 77491-620
2-Chloro-2,2-difluoro-1-(3-methyl-5-((3-methylbenzyl)oxy)phenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(3-methyl-5-((3-methylbenzyl)oxy)phenyl)ethanone

Precaución: For research use only

Formula: C₁₇H₁₅ClF₂O₂
MW: 324.7496 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892102
CAS Number: 2969459-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR