2,2,2-trifluoro-1-(3-methyl-5-((3-methylbenzyl)oxy)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

303767-1G 303767-500MG 303767-5G
77468-550EA 415.09 USD
77468-550 77481-234 77491-616
2,2,2-trifluoro-1-(3-methyl-5-((3-methylbenzyl)oxy)phenyl)ethanone ≥95%
2,2,2-trifluoro-1-(3-methyl-5-((3-methylbenzyl)oxy)phenyl)ethanone

Precaución: For research use only

Formula: C₁₇H₁₅F₃O₂
MW: 308.2950 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892100
CAS Number: 2969468-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR