1-(4-(diethylamino)phenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

303755-1G 303755-500MG 303755-5G
77468-546EA 415.09 USD
77468-546 77481-230 77491-612
1-(4-(diethylamino)phenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(4-(diethylamino)phenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₅H₂₅NO
Storage Temperature: Ambient
MDL Number: MFCD34892092
CAS Number: 2969328-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR