Ethyl 4-bromo-2,3-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

30354-1G 30354-500MG 30354-5G
77468-542EA 151.41 USD
77468-542 77481-226 77491-608
Ethyl 4-bromo-2,3-dimethylbenzoate ≥97%
Ethyl 4-bromo-2,3-dimethylbenzoate

Precaución: For research use only

Formula: C₁₁H₁₃BrO₂
MW: 257.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28738475
CAS Number: 1804403-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR