4-Bromo-1-isopropyl-2-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

30343-1G 30343-500MG 30343-5G
77468-538EA 339.9 USD
77468-538 77481-222 77491-604
4-Bromo-1-isopropyl-2-nitrobenzene ≥97%
4-Bromo-1-isopropyl-2-nitrobenzene

Precaución: For research use only

Formula: C₉H₁₀BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD22048623
CAS Number: 197711-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR