5-Chloro-2-ethoxypyridine-3-boronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

30325-1G 30325-500MG 30325-5G
77468-534EA 324.45 USD
77468-534 77481-218 77491-600
5-Chloro-2-ethoxypyridine-3-boronic acid pinacol ester ≥95%
5-Chloro-2-ethoxypyridine-3-boronic acid pinacol ester

Precaución: For research use only

Formula: C₁₃H₁₉BClNO₃
Storage Temperature: Ambient
MDL Number: MFCD16996801
CAS Number: 2121511-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR