Thiophene-2,3-dicarboxylic acid ≥95%

Supplier: AOBChem USA

Warning

30293-1G 30293-500MG 30293-5G
77468-530EA 106.09 USD
77468-530 77481-214 77491-596
Thiophene-2,3-dicarboxylic acid ≥95%
Thiophene-2,3-dicarboxylic acid

Precaución: For research use only

Formula: C₆H₄O₄S
Storage Temperature: Ambient
MDL Number: MFCD00034738
CAS Number: 1451-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR