2,2,3,3,3-pentafluoro-1-(3-isobutoxy-5-methylphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

302385-1G 302385-500MG 302385-5G
77468-518EA 415.09 USD
77468-518 77481-202 77491-582
2,2,3,3,3-pentafluoro-1-(3-isobutoxy-5-methylphenyl)propan-1-one ≥95%
2,2,3,3,3-pentafluoro-1-(3-isobutoxy-5-methylphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₄H₁₅F₅O₂
Storage Temperature: Ambient
MDL Number: MFCD34891732
CAS Number: 2969461-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR