2,2-Difluoro-1-(3-isobutoxy-5-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

302383-1G 302383-500MG 302383-5G
77468-514EA 415.09 USD
77468-514 77481-198 77491-578
2,2-Difluoro-1-(3-isobutoxy-5-methylphenyl)ethanone ≥95%
2,2-Difluoro-1-(3-isobutoxy-5-methylphenyl)ethanone

Precaución: For research use only

Formula: C₁₃H₁₆F₂O₂
MW: 242.2617 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34891730
CAS Number: 2969474-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR