Methyl(3-Methyl-4-nitrophenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

302244-1G 302244-500MG 302244-5G
77468-510EA 215.27 USD
77468-510 77481-194 77491-574
Methyl(3-Methyl-4-nitrophenyl)sulfane ≥95%
Methyl(3-Methyl-4-nitrophenyl)sulfane

Precaución: For research use only

Formula: C₈H₉NO₂S
Storage Temperature: Ambient
MDL Number: MFCD22489951
CAS Number: 308814-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR