1-Benzyloxy-2-bromo-4-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

30195-1G 30195-500MG 30195-5G
77468-506EA 273.98 USD
77468-506 77481-190 77491-570
1-Benzyloxy-2-bromo-4-(trifluoromethoxy)benzene ≥97%
1-Benzyloxy-2-bromo-4-(trifluoromethoxy)benzene

Precaución: For research use only

Formula: C₁₄H₁₀BrF₃O₂
MW: 347.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28973052
CAS Number: 143582-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR