1-Bromo-3-(2-chloroethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

300661-1G 300661-500MG 300661-5G
77468-502EA 109.18 USD
77468-502 77481-186 77491-564
1-Bromo-3-(2-chloroethoxy)benzene ≥95%
1-Bromo-3-(2-chloroethoxy)benzene

Precaución: For research use only

Formula: C₈H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD08276023
CAS Number: 730978-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR