1-Bromo-4-phenethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

300658-1G 300658-500MG 300658-5G
77468-498EA 363.59 USD
77468-498 77481-182 77491-560
1-Bromo-4-phenethoxybenzene ≥95%
1-Bromo-4-phenethoxybenzene

Precaución: For research use only

Formula: C₁₄H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD11123720
CAS Number: 30752-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR