2-Ethoxy-5-fluoroisonicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

30057-1G 30057-500MG 30057-5G
77468-494EA 224.54 USD
77468-494 77481-178 77491-556
2-Ethoxy-5-fluoroisonicotinaldehyde ≥97%
2-Ethoxy-5-fluoroisonicotinaldehyde

Precaución: For research use only

Formula: C₈H₈FNO₂
MW: 169.153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29036614
CAS Number: 1809157-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR