Cyclopropyl(5-ethyl-2-methoxyphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

300401-1G 300401-500MG 300401-5G
77468-490EA 347.11 USD
77468-490 77481-174 77491-552
Cyclopropyl(5-ethyl-2-methoxyphenyl)methanone ≥95%
Cyclopropyl(5-ethyl-2-methoxyphenyl)methanone

Precaución: For research use only

Formula: C₁₃H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD34891112
CAS Number: 1488906-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR