1-(2-(dimethylamino)phenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

299242-1G 299242-500MG 299242-5G
77468-478EA 260.59 USD
77468-478 77481-162 77491-536
1-(2-(dimethylamino)phenyl)-2,2,2-trifluoroethanone ≥95%
1-(2-(dimethylamino)phenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₀H₁₀F₃NO
CAS Number: 1256467-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR