N,N-Dimethyl-3-(tributylstannyl)aniline ≥95%

Supplier: AOBChem USA

Warning

299239-1G 299239-500MG 299239-5G
77468-474EA 415.09 USD
77468-474 77481-158 77491-532
N,N-Dimethyl-3-(tributylstannyl)aniline ≥95%
N,N-Dimethyl-3-(tributylstannyl)aniline

Precaución: For research use only

Formula: C₂₀H₃₇NSn
MW: 410.2244 g/mol
Storage Temperature: Ambient
CAS Number: 457644-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR