1-(3-(dimethylamino)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

299237-1G 299237-500MG 299237-5G
77468-470EA 415.09 USD
77468-470 77481-154 77491-528
1-(3-(dimethylamino)phenyl)cyclobutanol ≥95%
1-(3-(dimethylamino)phenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₂H₁₇NO
Storage Temperature: Ambient
CAS Number: 3002435-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR