2-Chloro-1-(3-(dimethylamino)phenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

299233-1G 299233-500MG 299233-5G
77468-466EA 515 USD
77468-466 77481-150 77491-524
2-Chloro-1-(3-(dimethylamino)phenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(3-(dimethylamino)phenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₁₀ClF₂NO
Storage Temperature: Ambient
CAS Number: 135334-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR