1-Bromo-2,5-difluoro-4-(2-propen-1-yl)benzene ≥95%

Supplier: AOBChem USA

Warning

298934-1G 298934-500MG 298934-5G
77468-462EA 310.03 USD
77468-462 77481-146 77491-520
1-Bromo-2,5-difluoro-4-(2-propen-1-yl)benzene ≥95%
1-Bromo-2,5-difluoro-4-(2-propen-1-yl)benzene

Precaución: For research use only

Formula: C₉H₇BrF₂
MW: 233.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847566
CAS Number: 1378861-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR