Ethyl 2-(4-Cyanophenyl)-2,2-difluoroacetate ≥95%

Supplier: AOBChem USA

Warning

298626-1G 298626-5G
77468-458EA 62.83 USD
77468-458 77491-516
Ethyl 2-(4-Cyanophenyl)-2,2-difluoroacetate ≥95%
Ethyl 2-(4-Cyanophenyl)-2,2-difluoroacetate

Precaución: For research use only

Formula: C₁₁H₉F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD25967876
CAS Number: 854778-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR