Trimethyl(3-methyl-5-propoxyphenyl)silane ≥95%

Supplier: AOBChem USA

Warning

297702-1G 297702-500MG 297702-5G
77468-454EA 415.09 USD
77468-454 77481-140 77491-512
Trimethyl(3-methyl-5-propoxyphenyl)silane ≥95%
Trimethyl(3-methyl-5-propoxyphenyl)silane

Precaución: For research use only

Formula: C₁₃H₂₂OSi
MW: 222.3986 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34890263
CAS Number: 2969434-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR