2,2,3,3,3-pentafluoro-1-(3-methyl-5-propoxyphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

297700-1G 297700-500MG 297700-5G
77468-450EA 415.09 USD
77468-450 77481-136 77491-508
2,2,3,3,3-pentafluoro-1-(3-methyl-5-propoxyphenyl)propan-1-one ≥95%
2,2,3,3,3-pentafluoro-1-(3-methyl-5-propoxyphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₃H₁₃F₅O₂
MW: 296.2331 g/mol
Storage Temperature: Ambient
CAS Number: 3002425-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR