2,2-Difluoro-1-(3-methyl-5-propoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

297698-1G 297698-500MG 297698-5G
77468-446EA 415.09 USD
77468-446 77481-132 77491-504
2,2-Difluoro-1-(3-methyl-5-propoxyphenyl)ethanone ≥95%
2,2-Difluoro-1-(3-methyl-5-propoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₄F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34890261
CAS Number: 2969326-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR