(4-Isopropoxy-3,5-dimethylphenyl)(isopropyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

297695-1G 297695-500MG 297695-5G
77468-442EA 415.09 USD
77468-442 77481-128 77491-500
(4-Isopropoxy-3,5-dimethylphenyl)(isopropyl)sulfane ≥95%
(4-Isopropoxy-3,5-dimethylphenyl)(isopropyl)sulfane

Precaución: For research use only

Formula: C₁₄H₂₂OS
MW: 238.3888 g/mol
Storage Temperature: Ambient
CAS Number: 3002507-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR