Ethyl(4-isopropoxy-3,5-dimethylphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

297689-1G 297689-500MG 297689-5G
77468-434EA 415.09 USD
77468-434 77481-120 77491-492
Ethyl(4-isopropoxy-3,5-dimethylphenyl)sulfane ≥95%
Ethyl(4-isopropoxy-3,5-dimethylphenyl)sulfane

Precaución: For research use only

Formula: C₁₃H₂₀OS
Storage Temperature: Ambient
CAS Number: 3002436-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR