Triethyl(4-isopropoxy-3,5-dimethylphenyl)silane ≥95%

Supplier: AOBChem USA

Warning

297687-1G 297687-500MG 297687-5G
77468-430EA 415.09 USD
77468-430 77481-116 77491-488
Triethyl(4-isopropoxy-3,5-dimethylphenyl)silane ≥95%
Triethyl(4-isopropoxy-3,5-dimethylphenyl)silane

Precaución: For research use only

Formula: C₁₇H₃₀OSi
MW: 278.505 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34890253
CAS Number: 2969463-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR