(4-Isopropoxy-3,5-dimethylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

297685-1G 297685-500MG 297685-5G
77468-426EA 415.09 USD
77468-426 77481-112 77491-484
(4-Isopropoxy-3,5-dimethylphenyl)(methyl)sulfane ≥95%
(4-Isopropoxy-3,5-dimethylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₂H₁₈OS
MW: 210.3357 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34890251
CAS Number: 2969245-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR