2-Chloro-2,2-difluoro-1-(4-isopropoxy-3,5-dimethylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

297683-1G 297683-500MG 297683-5G
77468-422EA 415.09 USD
77468-422 77481-108 77491-480
2-Chloro-2,2-difluoro-1-(4-isopropoxy-3,5-dimethylphenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(4-isopropoxy-3,5-dimethylphenyl)ethanone

Precaución: For research use only

Formula: C₁₃H₁₅ClF₂O₂
MW: 276.7068 g/mol
Storage Temperature: Ambient
CAS Number: 3002538-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR