2,2,2-trifluoro-1-(4-isopropoxy-3,5-dimethylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

297681-1G 297681-500MG 297681-5G
77468-418EA 415.09 USD
77468-418 77481-104 77491-476
2,2,2-trifluoro-1-(4-isopropoxy-3,5-dimethylphenyl)ethanone ≥95%
2,2,2-trifluoro-1-(4-isopropoxy-3,5-dimethylphenyl)ethanone

Precaución: For research use only

Formula: C₁₃H₁₅F₃O₂
MW: 260.2522 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34890249
CAS Number: 2969434-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR