1-(methoxymethoxy)-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

29704-1G 29704-500MG 29704-5G
77468-410EA 303.85 USD
77468-410 77481-096 77491-468
1-(methoxymethoxy)-3-methylbenzene ≥97%
1-(methoxymethoxy)-3-methylbenzene

Precaución: For research use only

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21195658
CAS Number: 57234-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR