Cyclopropyl(3,5-dimethyl-4-propoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

296453-1G 296453-500MG 296453-5G
77468-398EA 415.09 USD
77468-398 77481-084 77491-456
Cyclopropyl(3,5-dimethyl-4-propoxyphenyl)methanol ≥95%
Cyclopropyl(3,5-dimethyl-4-propoxyphenyl)methanol

Precaución: For research use only

Formula: C₁₅H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34889866
CAS Number: 2969398-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR