2-Chloro-1-(3,5-dimethyl-4-propoxyphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

296450-1G 296450-500MG 296450-5G
77468-394EA 415.09 USD
77468-394 77481-080 77491-452
2-Chloro-1-(3,5-dimethyl-4-propoxyphenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(3,5-dimethyl-4-propoxyphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₃H₁₅ClF₂O₂
MW: 276.7068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34889864
CAS Number: 2969298-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR