(4-(Benzyloxy)-2-methylphenyl)(isopropyl)sulfane ≥95%
Supplier: AOBChem USA
|
Warning
|
296446-1G
296446-500MG
296446-5G
77468-390EA
415.09
USD
77468-390
77481-076
77491-448
(4-(Benzyloxy)-2-methylphenyl)(isopropyl)sulfane ≥95%
(4-(Benzyloxy)-2-methylphenyl)(isopropyl)sulfane
Precaución: For research use only
Formula:
C₁₇H₂₀OS MW: 272.4051 g/mol Storage Temperature: Ambient |
CAS Number:
3002469-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...