(4-(Benzyloxy)-2-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

296442-1G 296442-500MG 296442-5G
77468-386EA 415.09 USD
77468-386 77481-072 77491-444
(4-(Benzyloxy)-2-methylphenyl)(phenyl)methanol ≥95%
(4-(Benzyloxy)-2-methylphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₂₁H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD34889858
CAS Number: 2685763-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR