1-(4-(Benzyloxy)-2-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

296439-1G 296439-500MG 296439-5G
77468-382EA 415.09 USD
77468-382 77481-068 77491-440
1-(4-(Benzyloxy)-2-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(4-(Benzyloxy)-2-methylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₉H₂₄O₂
MW: 284.3926 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34889857
CAS Number: 2969433-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR