(4-(Benzyloxy)-2-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

296436-1G 296436-500MG 296436-5G
77468-378EA 415.09 USD
77468-378 77481-064 77491-436
(4-(Benzyloxy)-2-methylphenyl)(methyl)sulfane ≥95%
(4-(Benzyloxy)-2-methylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₅H₁₆OS
MW: 244.3519 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34889854
CAS Number: 752133-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR