1-(4-(Benzyloxy)-2-methylphenyl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

296434-1G 296434-500MG 296434-5G
77468-374EA 594.31 USD
77468-374 77481-060 77491-432
1-(4-(Benzyloxy)-2-methylphenyl)-2-chloro-2,2-difluoroethanone ≥95%
1-(4-(Benzyloxy)-2-methylphenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₆H₁₃ClF₂O₂
Storage Temperature: Ambient
CAS Number: 1823777-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR