2-Bromo-4,6-dichloroanisole ≥97%

Supplier: AOBChem USA

Warning

29638-1G 29638-500MG 29638-5G
77468-370EA 348.14 USD
77468-370 77481-056 77491-428
2-Bromo-4,6-dichloroanisole ≥97%
2-Bromo-4,6-dichloroanisole

Precaución: For research use only

Formula: C₇H₅BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD08097451
CAS Number: 60633-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR