5-Bromo-3,4-difluoro-2-(trimethylsilyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

29582-1G 29582-500MG 29582-5G
77468-366EA 200.85 USD
77468-366 77481-052 77491-424
5-Bromo-3,4-difluoro-2-(trimethylsilyl)benzaldehyde ≥95%
5-Bromo-3,4-difluoro-2-(trimethylsilyl)benzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₁BrF₂OSi
MW: 293.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977322
CAS Number: 1809158-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR