2-butoxy-5-iodo-1,3-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

294803-1G 294803-500MG 294803-5G
77468-362EA 415.09 USD
77468-362 77481-048 77491-420
2-butoxy-5-iodo-1,3-dimethylbenzene ≥95%
2-butoxy-5-iodo-1,3-dimethylbenzene

Precaución: For research use only

Formula: C₁₂H₁₇IO
MW: 304.1672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34889508
CAS Number: 605666-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR