(4-butoxy-3,5-dimethylphenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

294801-1G 294801-500MG 294801-5G
77468-358EA 415.09 USD
77468-358 77481-044 77491-416
(4-butoxy-3,5-dimethylphenyl)triethylsilane ≥95%
(4-butoxy-3,5-dimethylphenyl)triethylsilane

Precaución: For research use only

Formula: C₁₈H₃₂OSi
MW: 292.5315 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34889507
CAS Number: 2969244-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR